Learn More
Biolegend FITC anti-mouse CD43
Supplier:  Biolegend 143204
Encompass_Preferred
FITC anti-mouse CD43 [S11]; Isotype: Rat IgG2b; Reactivity: Mouse; Apps: FC; Size: 200 ug
Catalog No. 50-165-291