Learn More
Biolegend FITC anti-mouse CD45.1
Supplier:  Biolegend 110706
Encompass_Preferred
FITC anti-mouse CD45.1 [A20]; Isotype: Mouse (A.SW) IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 500 ug
Catalog No. 50-163-420