Learn More
Biolegend FITC anti-mouse CD45 Recombinant
Supplier:  Biolegend 157608
Encompass_Preferred
FITC anti-mouse CD45 Recombinant [QA17A26]; Isotype: Mouse IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-207-2362