Learn More
Biolegend FITC anti-mouse CD49b
Supplier:  Biolegend 103503
Encompass_Preferred
FITC anti-mouse CD49b [HMα2]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-162-854