Learn More
Biolegend FITC anti-mouse CD69
Supplier:  Biolegend 104506
Encompass_Preferred
FITC anti-mouse CD69 [H1.2F3]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 500 ug
Catalog No. 50-162-938