Learn More
Biolegend FITC anti-mouse CD69
Supplier:  Biolegend 104505
Encompass_Preferred
FITC anti-mouse CD69 [H1.2F3]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-162-937