Learn More
Biolegend FITC anti-mouse CD83
Supplier:  Biolegend 121506
Encompass_Preferred
FITC anti-mouse CD83 [Michel-19]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-163-945