Learn More
Biolegend FITC anti-mouse CD8a
Supplier:  Biolegend 100706
Encompass_Preferred
FITC anti-mouse CD8a [53-6.7]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 500 ug
Catalog No. 50-162-477