Learn More
Biolegend FITC anti-mouse CD9
Supplier:  Biolegend 124807
Encompass_Preferred
FITC anti-mouse CD9 [MZ3]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-164-212