Learn More
Biolegend FITC anti-mouse CD94
Supplier:  Biolegend 105506
Encompass_Preferred
FITC anti-mouse CD94 [18d3]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 500 ug
Catalog No. 50-163-057