Learn More
Biolegend FITC anti-mouse I-Ek/rat RT1D
Supplier:  Biolegend 110205
Encompass_Preferred
FITC anti-mouse I-Ek/rat RT1D [14-4-4S]; Isotype: Mouse (C3H.SW) IgG2a, κ; Reactivity: Mouse, Cross-reacts with H-2d,p,r haplotypes. Mice of the H-2b,f,q,s haplotypes do not express I-E antigens. Cross-reacts with rat MHC class II antigen RT1D; Apps: FC; Size: 50 ug
Catalog No. 50-163-403