Learn More
Biolegend FITC anti-mouse IgG2a
Supplier:  Biolegend 407106
Encompass_Preferred
FITC anti-mouse IgG2a [RMG2a-62]; Isotype: Rat IgG; Reactivity: Mouse; Apps: FC; Size: 500 ug
Catalog No. 50-605-451