Learn More
Biolegend FITC anti-mouse Ly-51
Supplier:  Biolegend 108305
Encompass_Preferred
FITC anti-mouse Ly-51 [6C3]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-163-238