Learn More
Biolegend FITC anti-mouse Ly-6G
Supplier:  Biolegend 127605
Encompass_Preferred
FITC anti-mouse Ly-6G [1A8]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-164-389