Learn More
Biolegend FITC anti-mouse Ly49D
Supplier:  Biolegend 138303
Encompass_Preferred
FITC anti-mouse Ly49D [4E5]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-165-035