Learn More
Biolegend FITC anti-mouse/rat CD29
Supplier:  Biolegend 102206
Encompass_Preferred
FITC anti-mouse/rat CD29 [HMβ1-1]; Isotype: Armenian Hamster IgG; Reactivity: Mouse, Rat; Apps: FC; Size: 500 ug
Catalog No. 50-162-654