Learn More
Biolegend FITC anti-mouse TCR Vα3.2 [b, c]
Supplier:  Biolegend 135403
Encompass_Preferred
FITC anti-mouse TCR Vα3.2 [b, c] [RR3-16]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-164-842