Learn More
Biolegend FITC anti-mouse TCR Vβ7
Supplier:  Biolegend 118305
Encompass_Preferred
FITC anti-mouse TCR Vβ7 [TR310]; Isotype: Rat IgG2b, κ; Reactivity: Mouse TCR Vβ7 bearing T cells and NKT cells; may not be present in mice expressing the MIS-1a haplotype; Apps: FC; Size: 50 ug
Catalog No. 50-163-770