Learn More
Biolegend FITC anti-mouse TCR Vβ8.1, 8.2
Supplier:  Biolegend 118405
Encompass_Preferred
FITC anti-mouse TCR Vβ8.1, 8.2 [KJ16-133.18]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-163-775