Learn More
Biolegend FITC anti-rat CD71 [OX-26]; Isotype: Mouse IgG2a, ; Reactivity: Rat; Apps: FC; Size: 100 ug
Supplier:  Biolegend 204405
Encompass_Preferred
FITC anti-rat CD71 [OX-26]; Isotype: Mouse IgG2a, ; Reactivity: Rat; Apps: FC; Size: 100 ug
Catalog No. 50-165-455