Learn More
Biolegend FITC anti-rat IgG1 [MRG1-58]; Isotype: Mouse IgG; Reactivity: Rat; Apps: FC; Size: 500 ug
Supplier:  Biolegend 407406
Encompass_Preferred
FITC anti-rat IgG1 [MRG1-58]; Isotype: Mouse IgG; Reactivity: Rat; Apps: FC; Size: 500 ug
Catalog No. 50-169-204