Learn More
Sigma Aldrich Fine Chemicals Biosciences Albumin–fluorescein isothiocyanate conjugate, MFCD00282182, 100 mg
Catalog No. NC1080057
Encompass
Protein bovine, Synonym: FITC-albumin. Albumin−fluorescein isothiocyanate conjugate has been used: as a control to compare the intracellular delivery efficiency of proteins with or without carrier, for the fluorescent visualization of proteins and cells, as a blocking agent in ELISA, as a chemical for brain perfusion and fixation, for in vivo capillary–epithelial permeability experiment, and for confocal microscopy of right ventricular (RV) vasculature in rats.
Catalog No. NC1080057 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A9771100MG
Protein bovine, Synonym: FITC-albumin. Albumin−fluorescein isothiocyanate conjugate has been used: as a control to compare the intracellular delivery efficiency of proteins with or without carrier, for the fluorescent visualization of proteins and cells, as a blocking agent in ELISA, as a chemical for brain perfusion and fixation, for in vivo capillary–epithelial permeability experiment, and for confocal microscopy of right ventricular (RV) vasculature in rats.