Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BLD PHARMATECH CO LIMITED FITC, CAS Number: 3326-32-7, 1g, Purity: 95%

Catalog No. NC3965418
Encompass_Preferred

FITC, CAS Number: 3326-32-7, 1g, Purity: 95%

Catalog No. NC3965418 Supplier BLD PHARMATECH CO LIMITED Supplier No. BD12901G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

FITC, CAS Number: 3326-32-7, 1g, Purity: 95%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.