Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC FITC-conjugated Rabbit Anti-Human IgG H&L | 50 UL
SDP

Catalog No. 5000443707
Encompass_Preferred

FITC-conjugated Rabbit Anti-Human IgG H&L is a fluorescein isothiocyanate (FITC) labeled and rabbit-derived polyclonal antibody, which served as the secondary antibody.

  • Host: Rabbit
  • Immunogen: Native Human IgG
  • Conjugation: FITC
  • Modification: Unmodified
  • Isotype: IgG
  • Appearance: Liquid
  • Applications: ICC/IF, FC
  • Clonality: Polyclonal
  • Species reactivity: Human
  • Purity: Affinity purified
  • Storage & stability: Stored at -20°C for 1 year. Protect from light. Avoid repeated freeze / thaw cycles.

Catalog No. 50-004-43707 Supplier Medchemexpress LLC Supplier No. HYP8375250UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FITC-conjugated Rabbit Anti-Human IgG H&L is a fluorescein isothiocyanate (FITC) labeled and rabbit-derived polyclonal antibody, which served as the secondary antibody.

  • Host: Rabbit
  • Immunogen: Native Human IgG
  • Conjugation: FITC
  • Modification: Unmodified
  • Isotype: IgG
  • Appearance: Liquid
  • Applications: ICC/IF, FC
  • Clonality: Polyclonal
  • Species reactivity: Human
  • Purity: Affinity purified
  • Storage & stability: Stored at -20°C for 1 year. Protect from light. Avoid repeated freeze / thaw cycles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.