Learn More
Abcam FITC Conjugation Kit (Fast) - Lightning-Link, 100UG
Catalog No. NC1840279
Encompass_Preferred
The Abcam FITC Conjugation Kit (ab188285) offers: -Speed and Efficiency: enables FITC conjugation or labeling in less than 20 minutes with only 30 seconds of hands-on time. -High Recovery: ensures 100% antibody recovery, meaning no loss of valuable antibodies. -Versatility: can be used to conjugate not only antibodies but also other proteins and peptides, making it suitable for various applications - Confidence: cited in over 15 publications
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1840279 Supplier Abcam Supplier No. AB188285100UG
The Abcam FITC Conjugation Kit (ab188285) offers: -Speed and Efficiency: enables FITC conjugation or labeling in less than 20 minutes with only 30 seconds of hands-on time. -High Recovery: ensures 100% antibody recovery, meaning no loss of valuable antibodies. -Versatility: can be used to conjugate not only antibodies but also other proteins and peptides, making it suitable for various applications - Confidence: cited in over 15 publications
The product is subject to the following: Abcam Restricted Use Statement