Learn More
Abcam FITC Conjugation Kit - Lightning-Link, 1MG
Catalog No. 502585605
Encompass_Preferred
Abcam's FITC Conjugation Kit offers ab102884: - Rapid Conjugation: achieve FITC labeling in under 4 hours with just 30 seconds of hands-on time. - High Efficiency: ensures 100% antibody recovery, preserving the integrity and value of your antibodies. - Versatility: suitable for conjugating antibodies, proteins, and peptides, and the labeled antibodies can be used immediately in applications such as Western Blotting (WB), ELISA, and Immunohistochemistry (IHC) without further purification. - Confidence: cited in over 30 publications
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-5605 Supplier Abcam Supplier No. AB1028841MG
Abcam's FITC Conjugation Kit offers ab102884: - Rapid Conjugation: achieve FITC labeling in under 4 hours with just 30 seconds of hands-on time. - High Efficiency: ensures 100% antibody recovery, preserving the integrity and value of your antibodies. - Versatility: suitable for conjugating antibodies, proteins, and peptides, and the labeled antibodies can be used immediately in applications such as Western Blotting (WB), ELISA, and Immunohistochemistry (IHC) without further purification. - Confidence: cited in over 30 publications
The product is subject to the following: Abcam Restricted Use Statement