Learn More
Chondrex Inc FITC-DEXTRAN, 125 MG, 40 KDA
Supplier:  Chondrex Inc 4009
Encompass_Preferred
FITC-Dextran (40 kDa) solution, 25 mg/mL x 5 mL, in PBS (green color). Fluorescent Properties: Excitation - 490 nm, Emission - 520 nm.
Catalog No. 50-152-6773