Learn More
Medchemexpress LLC HY-128868 5mg Medchemexpress, FITC-Dextran CAS:60842-46-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1288685MG
Encompass_Preferred
Medchemexpress, HY-128868 5mg FITC-Dextran CAS:60842-46-8 FITC-Dextran is a marker consisting of coupling fluorescein-isothiocyanate to dextran. FITC-Dextrans are polysaccharides comprised of varying lengths of branched glucose molecules and can be used to determine solute, ion and protein permeability of the blood-brain barrier (BBB) based on the size of the dextran used [1] [2] [3] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2642