Learn More
Sigma Aldrich Fine Chemicals Biosciences Fluorescein isothiocyanate–dextran, 60842-46-8, MFCD00131092, 100 mg
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 46945100MGF
Encompass
Average mol wt 70,000, (FITC:Glucose = 1:250), Synonym: FITC–Dextran, FITC-Dextran 70. Fluorescein isothiocyanate–dextran (FITC-Dextran 70) is use as a fluorescent probe to study cell processes such as cell permeability, phagocytosis and endocytosis and to study mechanisms of biomolecular delivery. FITC-Dextran 70 may be used to study the capacity, selectivity and efficiency of encapsulation and release by drug delivery vehicles.
Catalog No. NC0689651