Learn More
Apexbio Technology LLC FITC-Dextran (MW 70000) 5mg
SDP
Catalog No. 5000093371
Encompass_Preferred
FITC-Dextran MW 70000 (CAS 60842-46-8) is a fluorescent-labeled polysaccharide probe synthesized by conjugating fluorescein isothiocyanate (FITC) to dextran It is designed to enable visualization and quantitative analysis of permeability endocytic uptake and transport phenomena in biological systems FITC-Dextran MW 70000 exhibits its utility by emitting fluorescence upon excitation (Ex 495 nm Em 519 nm) allowing detection via fluorescence microscopy flow cytometry or spectrofluorometry In experimental models it is commonly used for evaluating epithelial barrier integrity in intestinal vascular and blood-brain barrier assays and for investigating uptake mechanisms in cellular biology Based on these properties FITC-Dextran MW 70000 is valuable for research applications in membrane permeability trans-barrier transport studies endocytosis assays and cellular uptake investigations
Catalog No. 50-000-93371 Supplier Apexbio Technology LLC Supplier No. A90655
FITC-Dextran MW 70000 (CAS 60842-46-8) is a fluorescent-labeled polysaccharide probe synthesized by conjugating fluorescein isothiocyanate (FITC) to dextran It is designed to enable visualization and quantitative analysis of permeability endocytic uptake and transport phenomena in biological systems FITC-Dextran MW 70000 exhibits its utility by emitting fluorescence upon excitation (Ex 495 nm Em 519 nm) allowing detection via fluorescence microscopy flow cytometry or spectrofluorometry In experimental models it is commonly used for evaluating epithelial barrier integrity in intestinal vascular and blood-brain barrier assays and for investigating uptake mechanisms in cellular biology Based on these properties FITC-Dextran MW 70000 is valuable for research applications in membrane permeability trans-barrier transport studies endocytosis assays and cellular uptake investigations