Learn More
Medchemexpress LLC FITC-dextran sulfate | 10 MG
SDP
Catalog No. 5000295113
Encompass_Preferred
FITC-dextran sulfate (MW 500 kDa) is a high-molecular-weight, anionic polysaccharide covalently labeled with fluorescein isothiocyanate (FITC). It is used as a fluorescent tracer and permeability probe in biological and fluorescence-based assays where a large, anionic, fluorescent marker is required. Handle and store protected from light at low temperatures for best stability.
  • Fluorescently labeled with FITC for robust fluorescence signal.
  • High average molecular weight (~500 kDa) for restricted diffusion and barrier permeability studies.
  • Anionic dextran sulfate backbone for charge-based interactions.
  • Store protected from light at low temperatures to preserve stability.
  • Offered in small-scale quantities suitable for laboratory experiments.
Catalog No. 50-002-95113 Supplier Medchemexpress LLC Supplier No. HY172163C10MG
FITC-dextran sulfate (MW 500 kDa) is a high-molecular-weight, anionic polysaccharide covalently labeled with fluorescein isothiocyanate (FITC). It is used as a fluorescent tracer and permeability probe in biological and fluorescence-based assays where a large, anionic, fluorescent marker is required. Handle and store protected from light at low temperatures for best stability.
  • Fluorescently labeled with FITC for robust fluorescence signal.
  • High average molecular weight (~500 kDa) for restricted diffusion and barrier permeability studies.
  • Anionic dextran sulfate backbone for charge-based interactions.
  • Store protected from light at low temperatures to preserve stability.
  • Offered in small-scale quantities suitable for laboratory experiments.