Learn More
Medchemexpress LLC FITC-dextran sulfate | 3,769.00 Da | 10 MG
SDP
Catalog No. 5000295097
Encompass_Preferred
FITC-dextran sulfate (MW 4 kDa) is a fluorescein-labeled sodium dextran sulfate (average molecular weight ≈4 kDa) supplied as a light yellow to yellow solid for research use. It is water-soluble and commonly used as a fluorescent tracer or probe in biological and biochemical assays.
  • Fluorescently labeled with FITC for visualization in fluorescence assays.
  • Average molecular weight ≈ 3,769 Da (reported ~4,000 Da).
  • Degree of substitution 0.005.
  • Light yellow to yellow solid for easy handling.
  • Storage: protect from light; solid -20°C; in solvent -80°C (6 months) or -20°C (1 month).
  • Loss on drying 1.20% (per COA).
  • For research use only.
Catalog No. 50-002-95097 Supplier Medchemexpress LLC Supplier No. HY17216310MG
FITC-dextran sulfate (MW 4 kDa) is a fluorescein-labeled sodium dextran sulfate (average molecular weight ≈4 kDa) supplied as a light yellow to yellow solid for research use. It is water-soluble and commonly used as a fluorescent tracer or probe in biological and biochemical assays.
  • Fluorescently labeled with FITC for visualization in fluorescence assays.
  • Average molecular weight ≈ 3,769 Da (reported ~4,000 Da).
  • Degree of substitution 0.005.
  • Light yellow to yellow solid for easy handling.
  • Storage: protect from light; solid -20°C; in solvent -80°C (6 months) or -20°C (1 month).
  • Loss on drying 1.20% (per COA).
  • For research use only.