Learn More
Medchemexpress LLC FITC-dextran sulfate | 4000.00 | 25 MG
SDP
Catalog No. 5000385140
Encompass_Preferred
FITC-dextran sulfate, with a molecular weight of 4 kDa, is a sodium dextran sulfate that has been labeled with Fluorescein Isothiocyanate (FITC). This product is intended for research purposes only.
  • Labeled with FITC for fluorescent detection.
  • Average molecular weight of 4 kDa.
  • For research use only.
  • Not suitable for patient use.
Catalog No. 50-003-85140 Supplier Medchemexpress LLC Supplier No. 25MGHY172163C
FITC-dextran sulfate, with a molecular weight of 4 kDa, is a sodium dextran sulfate that has been labeled with Fluorescein Isothiocyanate (FITC). This product is intended for research purposes only.
  • Labeled with FITC for fluorescent detection.
  • Average molecular weight of 4 kDa.
  • For research use only.
  • Not suitable for patient use.