Learn More
Medchemexpress LLC FITC-dextran sulfate | 500,000 | 5 MG
SDP
Catalog No. 5000386659
Encompass_Preferred
FITC-dextran sulfate, with an average molecular weight of 500 kDa, is a sodium dextran sulfate labeled with FITC. This product is intended for research use only.
  • Labeled with FITC
  • Average molecular weight of 500 kDa
  • Suitable for research applications
Catalog No. 50-003-86659 Supplier Medchemexpress LLC Supplier No. HY172163C5MG
FITC-dextran sulfate, with an average molecular weight of 500 kDa, is a sodium dextran sulfate labeled with FITC. This product is intended for research use only.
  • Labeled with FITC
  • Average molecular weight of 500 kDa
  • Suitable for research applications