Learn More
Medchemexpress LLC FITC-dextran sulfate (MW 4kDa) | 4000.00 | 100 MG
SDP
Catalog No. 5000386462
Encompass_Preferred
FITC-dextran sulfate, 4kDa is a sodium dextran sulfate labeled with FITC, with an average molecular weight of 4 kDa.
  • Biochemical assay reagents
  • Cell assay reagents
  • Inflammation or immune system disease reagents
  • Fluorescent dye
Catalog No. 50-003-86462 Supplier Medchemexpress LLC Supplier No. HY172163100MG
FITC-dextran sulfate, 4kDa is a sodium dextran sulfate labeled with FITC, with an average molecular weight of 4 kDa.
  • Biochemical assay reagents
  • Cell assay reagents
  • Inflammation or immune system disease reagents
  • Fluorescent dye