Learn More
Medchemexpress LLC FITC-dextran sulfate (MW 4kDa) | 4000.00 | 5 MG
SDP
Catalog No. 5000386165
Encompass_Preferred
FITC-dextran sulfate, 4kDa is a sodium dextran sulfate labeled with FITC, with an average molecular weight of 4 kDa. It is for research use only.
  • Fluorescent dye
Catalog No. 50-003-86165 Supplier Medchemexpress LLC Supplier No. HY1721635MG
FITC-dextran sulfate, 4kDa is a sodium dextran sulfate labeled with FITC, with an average molecular weight of 4 kDa. It is for research use only.
  • Fluorescent dye