Learn More
Medchemexpress LLC FITC-dextran sulfate (MW 4kDa) | 4000.00 | 50 MG
SDP
Catalog No. 5000386600
Encompass_Preferred
FITC-dextran sulfate, 4 kDa, is a sodium dextran sulfate labeled with FITC. It has an average molecular weight of 4 kDa and is intended for research use only.
  • Fluorescent dye
  • Used as a biochemical assay reagent
  • Used as a cell assay reagent
Catalog No. 50-003-86600 Supplier Medchemexpress LLC Supplier No. HY17216350MG
FITC-dextran sulfate, 4 kDa, is a sodium dextran sulfate labeled with FITC. It has an average molecular weight of 4 kDa and is intended for research use only.
  • Fluorescent dye
  • Used as a biochemical assay reagent
  • Used as a cell assay reagent