Learn More
Medchemexpress LLC Fitc-dextran sulfate (mw 500kda) | 500000 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY172163C
Encompass_Preferred
FITC-dextran sulfate, 500kDa is a sodium dextran sulfate labeled with FITC, with an average molecular weight of 500 kDa. It is for research use only.
  • Sodium dextran sulfate labeled with FITC
  • Average molecular weight of 500 kDa
  • For research use only
Catalog No. 50-003-85545