Learn More
Diagnostic Biosystems FITC DILUENT FITC Diluent
Catalog No. 502407347
Encompass_Preferred
This product can be used as a diluent to prepare primary antibodies conjugated to FITC. It can be used with any primary antibody irrespective of its source of origin. FITC Diluent is specifically designed to stabilize primary antibodies and reduce nonspecific background staining observed during immunostaining.\n\nIn order to obtain optimal staining, correct dilution as well as selection of appropriate diluent is critical. Ionic strength, pH of the environment and the presence or absence of detergents can affect the binding as well as the stability of the primary antibody. The optimal pH together with stabilizing proteins in FITC Diluent ensures the maximum stability of primary antibodies. Our FITC Diluent contains optimal concentrations of sodium azide and detergents to prevent nonspecific binding and enable better penetration of primary antibodies. FITC Diluent is specifically designed for diluting primary antibodies for immunohistochemistry.
Catalog No. 50-240-7347 Supplier Diagnostic Biosystems Supplier No. FD001100
This product can be used as a diluent to prepare primary antibodies conjugated to FITC. It can be used with any primary antibody irrespective of its source of origin. FITC Diluent is specifically designed to stabilize primary antibodies and reduce nonspecific background staining observed during immunostaining.\n\nIn order to obtain optimal staining, correct dilution as well as selection of appropriate diluent is critical. Ionic strength, pH of the environment and the presence or absence of detergents can affect the binding as well as the stability of the primary antibody. The optimal pH together with stabilizing proteins in FITC Diluent ensures the maximum stability of primary antibodies. Our FITC Diluent contains optimal concentrations of sodium azide and detergents to prevent nonspecific binding and enable better penetration of primary antibodies. FITC Diluent is specifically designed for diluting primary antibodies for immunohistochemistry.