Learn More
Cytek Biosciences Inc FITC aHu CD45RA Anti Primary
Supplier: Cytek Biosciences Inc 350458T100
Product Name: FITC Anti-Human CD45RA (HI100)
Product Class: Antibody
Type: Primary
Product Name: FITC Anti-Human CD45RA (HI100)
Cytek Catalog Number: 35-0458-T100
Marker: CD45RA
Target Species: Human
Clonality: Monoclonal
Clone: HI100
Host Species: Mouse
Isotype: IgG2b, k
Conjugation: FITC
Size: 100 Tests
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.