Learn More
Sigma Aldrich Fine Chemicals Biosciences Insulin-FITC labeled human, 5 mg
Supplier:  Sigma Aldrich Fine Chemicals Biosciences I36615MG
Encompass
Recombinant, expressed in yeast, lyophilized powder, storage temp: −20°C. Insulin-FITC labeled human has been used in primary endothelial cells from human adipose tissue microvasculature (HAMEC) to study insulin transcytosis.
Catalog No. NC1080387