Learn More
Biotium FITC (Isomer I)
Supplier:  Biotium 90020
Encompass_Preferred
FITC (fluorescein-5-isothiocyanate) is an amine-reactive fluorescein derivative for labeling proteins or other molecules containing primary amines. Properties: Ex / Em = 492/520 nm; e = 75,000; Orange solid soluble in DMF or DMSO; Store at -20°C and protect from light, especially in solution; C 21 H 11 NO 5 S; MW: 389.4; [3326-32-7]
Catalog No. 50-196-4803