Learn More
Medchemexpress LLC FITC-Labeled CD70 H 50ug | 50ug
SDP
Catalog No. 5000194475
Encompass_Preferred
CD70 is the ligand of CD27 in activated T and B lymphocytes is an important cytokine in CD70-CD27 pathway involving with the generation and maintenance of T cell immunity[1] CD70 is a surface antigen also shows antiviral activity and regulates viability of tumor cells and regulatory T cells[2][3] As for CD70 protein in human contains transmembrane domain and transmembrane helix belonging to the tumor necrosis factor (TNF) family CD70 Protein Human (FITC HEK293 His) is a luorescein isothiocyanate (FITC)-labeled recombinant protein expressed in the HEK293 cells with a N terminal His-tag
Catalog No. 50-001-94475 Supplier Medchemexpress LLC Supplier No. HYP7854050UG
CD70 is the ligand of CD27 in activated T and B lymphocytes is an important cytokine in CD70-CD27 pathway involving with the generation and maintenance of T cell immunity[1] CD70 is a surface antigen also shows antiviral activity and regulates viability of tumor cells and regulatory T cells[2][3] As for CD70 protein in human contains transmembrane domain and transmembrane helix belonging to the tumor necrosis factor (TNF) family CD70 Protein Human (FITC HEK293 His) is a luorescein isothiocyanate (FITC)-labeled recombinant protein expressed in the HEK293 cells with a N terminal His-tag