Learn More
ACROBiosystems ACROBiosystems FITC-Labeled Human Angiopoietin-2 / ANGPT2 (275-496) Protein, His Tag
Catalog No. 502106489
Encompass_Preferred
Recombinant Protein;200UG;FITC-Labeled Human Angiopoietin-2 (275-496), His Tag (AN2-HF242) is expressed from human 293 cells (HEK293). It contains AA Lys 275 - Phe 496 (Accession # AAI26201.1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 26.3 kDa. The protein migrates as 30-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6489 Supplier ACROBiosystems Supplier No. AN2HF242200UG
Recombinant Protein;200UG;FITC-Labeled Human Angiopoietin-2 (275-496), His Tag (AN2-HF242) is expressed from human 293 cells (HEK293). It contains AA Lys 275 - Phe 496 (Accession # AAI26201.1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 26.3 kDa. The protein migrates as 30-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.