Recombinant Protein;25UG;FITC-Labeled Human CD27 Ligand, His,Flag Tag (CDD-HF2D4) is expressed from human 293 cells (HEK293). It contains AA Ser 52 - Pro 193 (Accession # P32970-1). It is the biotinylated form of Human CD27 Ligand, His,Flag Tag (active trimer) (MALS verified) (Cat. No.CDL-H52Da).;This protein carries a polyhistidine tag at the N-terminus, followed by a flag tag. The protein has a calculated MW of 49.3 kDa. The protein migrates as 50-55 kDa under non-reducing (NR) condition (SDS-PAGE) due to glycosylation.;Measured by its binding ability in a functional ELISA. Immobilized Anti-CD27 Ligand Antibody, Human IgG1 at 1 μg/mL (100 μL/well) can bind FITC-Labeled Human CD27 Ligand, His,Flag Tag (Cat. No. CDD-HF2D4) with a linear range of 0.039-0.156 μg/mL (QC tested).;Measured by its binding ability in a functional ELISA. Immobilized Human CD27, Fc Tag (Cat. No. CD7-H5254) at 5 μg/mL (100 μL/well) can bind FITC-Labeled Human CD27 Ligand, His,Flag Tag (Cat. No. CDD-HF2D4) with a