Learn More
ACROBiosystems ACROBiosystems FITC-Labeled Human CD30 / TNFRSF8 Protein, His Tag
Catalog No. 502106294
Encompass_Preferred
Recombinant Protein;200UG;FITC-Labeled Human CD30, His Tag (CD0-HF2H4) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Lys 379 (Accession # NP_001234.2).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 40.4 kDa. The protein migrates as 60-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Monoclonal Anti-Human CD30 Human Antibody, Human IgG1 at 2 μg/mL (100 μL/well) can bind FITC-Labeled Human CD30, His Tag (Cat. No. CD0-HF2H4) with a linear range of 10-156 ng/mL (QC tested).;2e5 of anti-CD30 CAR-293 cells were stained with 100 μL of 1 μg/mL of FITC-Labeled Human CD30, His Tag (Cat. No.CD0-HF2H4) and negative control protein respectively, FITC signal was used to evaluate the binding activity (QC tested).
Catalog No. 50-210-6294 Supplier ACROBiosystems Supplier No. CD0HF2H4200UG
Add to Cart
Add to Cart
Recombinant Protein;200UG;FITC-Labeled Human CD30, His Tag (CD0-HF2H4) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Lys 379 (Accession # NP_001234.2).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 40.4 kDa. The protein migrates as 60-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Monoclonal Anti-Human CD30 Human Antibody, Human IgG1 at 2 μg/mL (100 μL/well) can bind FITC-Labeled Human CD30, His Tag (Cat. No. CD0-HF2H4) with a linear range of 10-156 ng/mL (QC tested).;2e5 of anti-CD30 CAR-293 cells were stained with 100 μL of 1 μg/mL of FITC-Labeled Human CD30, His Tag (Cat. No.CD0-HF2H4) and negative control protein respectively, FITC signal was used to evaluate the binding activity (QC tested).