Learn More
ACROBiosystems ACROBiosystems FITC-Labeled Human CD37 Protein, Fc Tag
Catalog No. 502106479
Encompass_Preferred
Recombinant Protein;25UG;FITC-Labeled Human CD37, Fc Tag (CD7-HF26x) is expressed from human 293 cells (HEK293). It contains AA Arg 112 - Asn 241 (Accession # P11049-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 41.3 kDa. The protein migrates as 50-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6479 Supplier ACROBiosystems Supplier No. CD7HF26X25UG
Recombinant Protein;25UG;FITC-Labeled Human CD37, Fc Tag (CD7-HF26x) is expressed from human 293 cells (HEK293). It contains AA Arg 112 - Asn 241 (Accession # P11049-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 41.3 kDa. The protein migrates as 50-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.