Learn More
ACROBiosystems FITC-Labeled Human CD38 Protein, His Tag
Supplier:  ACROBiosystems CD8HF2H5200UG
Encompass_Preferred
Recombinant Protein;200UG;FITC-Labeled Human CD38, His Tag (Cat. No. CD8-HF2H5) is expressed from human HEK293 cells. It contains AA Val 43 - Ile 300 (Accession # NP_001766). It is the FITC labeled form of Human CD38 Protein, His Tag (Cat. No. CD8-H5224).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 30.7 kDa. The protein migrates as 40-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-CD38 Antibody (Daratumumab), Human IgG1 at 2 μg/mL (100 μL/well) can bind FITC-Labeled Human CD38, His Tag (Cat. No. CD8-HF2H5) with a linear range of 1.25-10 μg/mL (QC tested).
Catalog No. 50-205-5173