Learn More
ACROBiosystems FITC-Labeled Human CD5 Protein, His Tag (MALS verified)
Catalog No. 502055170
Encompass_Preferred
Recombinant Protein;25UG;FITC-Labeled Human CD5, His Tag (CD5-H52H2) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Pro 372 (Accession # P06127-1). It is the FITC labeled form of Human CD5, His Tag (CD5-H52H5).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 40.5 kDa. The protein migrates as 45-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-205-5170 Supplier ACROBiosystems Supplier No. CD5H52H225UG
Recombinant Protein;25UG;FITC-Labeled Human CD5, His Tag (CD5-H52H2) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Pro 372 (Accession # P06127-1). It is the FITC labeled form of Human CD5, His Tag (CD5-H52H5).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 40.5 kDa. The protein migrates as 45-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.