Learn More
ACROBiosystems ACROBiosystems FITC-Labeled Human CD7 Protein, Fc Tag
Catalog No. 502106881
Encompass_Preferred
Recombinant Protein;25UG;FITC-Labeled Human CD7, Fc Tag (CD7-HF258) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Pro 180 (Accession # P09564-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 42.9 kDa.
Catalog No. 50-210-6881 Supplier ACROBiosystems Supplier No. CD7HF25825UG
Recombinant Protein;25UG;FITC-Labeled Human CD7, Fc Tag (CD7-HF258) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Pro 180 (Accession # P09564-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 42.9 kDa.